Assay Details
Gene Symbol: Ckmt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198415.2 | NP_940807.1 | 1 - 2 | 62 | 71 | |
GenBank mRNA | BC061221.1 | - | 1 - 2 | 62 | 30296538 | 71 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC061221.1, NM_198415.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
