Assay Details
Gene Symbol: Ckmt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198415.2 | NP_940807.1 | 4 - 5 | 522 | 82 | |
GenBank mRNA | AK009042.1 | - | 2 - 3 | 226 | 82 | |
BC061221.1 | - | 4 - 5 | 522 | 30296538 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
