Assay Details
| Gene Symbol: Ckmt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198415.2 | NP_940807.1 | 7 - 8 | 965 | 86 | |
| GenBank mRNA | AK009042.1 | - | 5 - 6 | 669 | 86 | |
| BC061221.1 | - | 7 - 8 | 965 | 30296538 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ckmt2
Genomic Map