| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0100024701.mm.1 |
NM_177129.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100024701.mm.1 |
NM_177129.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100024701.mm.1 |
TC0100003079.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100024702.mm.1 |
NM_177129.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100024702.mm.1 |
NM_177129.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100024702.mm.1 |
TC0100003079.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100024700.mm.1 |
NM_177129.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100024700.mm.1 |
NM_177129.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100024700.mm.1 |
TC0100003079.mm.1 |
MTA TC |