Assay Details
| Gene Symbol: Dstyk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172516.4 | NP_766104.2 | 3 - 4 | 1468 | 69 | |
| GenBank mRNA | AK083696.1 | - | 3 - 4 | 1366 | 69 | |
| AK132119.1 | - | 3 - 4 | 1343 | 69 | ||
| AK145449.1 | - | 3 - 4 | 1313 | 69 | ||
| AK172954.1 | - | 3 - 4 | 1314 | 69 | ||
| AK180137.1 | - | 1 - 2 | 178 | 69 | ||
| AK210916.1 | - | 1 - 2 | 314 | 69 | ||
| AY208852.1 | - | 3 - 4 | 1315 | 69 | ||
| AY429676.1 | - | 3 - 4 | 1354 | 69 | ||
| BC019133.1 | - | 1 - 2 | 103 | 69 | ||
| BC130226.1 | - | 3 - 4 | 1344 | 40141770 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dstyk
Genomic Map