Assay Details
| Gene Symbol: Dstyk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172516.4 | NP_766104.2 | 4 - 5 | 1706 | 105 | |
| GenBank mRNA | AK083696.1 | - | 4 - 5 | 1604 | 105 | |
| AK132119.1 | - | 4 - 5 | 1581 | 105 | ||
| AK145449.1 | - | 4 - 5 | 1551 | 105 | ||
| AK172954.1 | - | 4 - 5 | 1552 | 105 | ||
| AY208852.1 | - | 4 - 5 | 1553 | 105 | ||
| AY429676.1 | - | 4 - 5 | 1592 | 105 | ||
| BC019133.1 | - | 2 - 3 | 341 | 105 | ||
| BC089380.1 | - | 1 - 2 | 123 | 105 | ||
| BC130226.1 | - | 4 - 5 | 1582 | 40141770 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dstyk
Genomic Map