Assay Details
| Gene Symbol: Dstyk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172516.4 | NP_766104.2 | 5 - 6 | 1793 | 70 | |
| GenBank mRNA | AK050542.1 | - | 1 - 2 | 39 | 70 | |
| AK083696.1 | - | 5 - 6 | 1691 | 70 | ||
| AK132119.1 | - | 5 - 6 | 1668 | 70 | ||
| AK145449.1 | - | 5 - 6 | 1638 | 70 | ||
| AK172954.1 | - | 5 - 6 | 1639 | 70 | ||
| AY208852.1 | - | 5 - 6 | 1640 | 70 | ||
| AY429676.1 | - | 5 - 6 | 1679 | 70 | ||
| BC019133.1 | - | 3 - 4 | 428 | 70 | ||
| BC089380.1 | - | 2 - 3 | 210 | 70 | ||
| BC130226.1 | - | 5 - 6 | 1669 | 40141770 | 70 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK050542.1 transcripts.
Set Membership: |
|

Dstyk
Genomic Map