Assay Details
Gene Symbol: Gm12169 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163357.1 | NP_001156829.1 | 3 - 4 | 506 | 68 | |
GenBank mRNA | BG066735.2 | - | 4 - 5 | 68 | ||
BG079892.2 | - | 3 - 4 | 491 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
