Assay Details
| Gene Symbol: Scly | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016717.3 | NP_057926.2 | 2 - 3 | 228 | 63 | |
| GenBank mRNA | AF175407.1 | - | 2 - 3 | 180 | 63 | |
| AK036533.1 | - | 2 - 3 | 197 | 63 | ||
| AK036750.1 | - | 2 - 3 | 230 | 63 | ||
| AK178608.1 | - | 2 - 3 | 163 | 63 | ||
| AK183254.1 | - | 2 - 3 | 163 | 63 | ||
| AK188684.1 | - | 2 - 3 | 178 | 63 | ||
| BC019879.1 | - | 2 - 3 | 188 | 5123843 | 63 | |
| BC021389.1 | - | 2 - 3 | 212 | 5054691 | 63 | |
More Information
Set Membership: |
|

Scly
Genomic Map