Assay Details
| Gene Symbol: Scly | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016717.3 | NP_057926.2 | 4 - 5 | 510 | 65 | |
| GenBank mRNA | AF175407.1 | - | 4 - 5 | 462 | 65 | |
| AK036750.1 | - | 4 - 5 | 512 | 65 | ||
| BC019879.1 | - | 4 - 5 | 470 | 5123843 | 65 | |
| BC021389.1 | - | 4 - 5 | 494 | 5054691 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Scly
Genomic Map