Assay Details
| Gene Symbol: Scly | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016717.3 | NP_057926.2 | 6 - 7 | 791 | 80 | |
| GenBank mRNA | AF175407.1 | - | 6 - 7 | 743 | 80 | |
| AK036533.1 | - | 6 - 7 | 760 | 80 | ||
| AK036750.1 | - | 6 - 7 | 793 | 80 | ||
| BC019879.1 | - | 6 - 7 | 751 | 5123843 | 80 | |
| BC021389.1 | - | 6 - 7 | 775 | 5054691 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Scly
Genomic Map