Assay Details
| Gene Symbol: Scly | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016717.3 | NP_057926.2 | 7 - 8 | 905 | 65 | |
| GenBank mRNA | AF175407.1 | - | 7 - 8 | 857 | 65 | |
| AK036750.1 | - | 7 - 8 | 907 | 65 | ||
| AK206313.1 | - | 1 - 2 | 90 | 65 | ||
| BC019879.1 | - | 7 - 8 | 865 | 5123843 | 65 | |
| BC021389.1 | - | 7 - 8 | 889 | 5054691 | 65 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Scly
Genomic Map