Assay Details
| Gene Symbol: Ilkap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023343.2 | NP_075832.1 | 8 - 9 | 793 | 72 | |
| GenBank mRNA | AF117831.1 | - | 3 - 4 | 247 | 72 | |
| AK003007.1 | - | 7 - 8 | 1766 | 72 | ||
| AK005408.1 | - | 8 - 9 | 816 | 72 | ||
| AK019158.1 | - | 2 - 3 | 177 | 72 | ||
| BC011427.1 | - | 8 - 9 | 763 | 72 | ||
| BC026953.1 | - | 8 - 9 | 793 | 5133980 | 72 | |
| BC027439.1 | - | 8 - 9 | 682 | 4164301 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ilkap
Genomic Map