Assay Details
| Gene Symbol: Egln3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028133.2 | NP_082409.2 | 2 - 3 | 787 | 106 | |
| GenBank mRNA | AJ310548.1 | - | 2 - 3 | 768 | 106 | |
| AK011502.1 | - | 1 - 2 | 91 | 106 | ||
| AK044787.1 | - | 2 - 3 | 789 | 106 | ||
| AK165972.1 | - | 2 - 3 | 718 | 106 | ||
| AK170732.1 | - | 2 - 3 | 718 | 106 | ||
| BC044926.1 | - | 2 - 3 | 697 | 5373240 | 106 | |
| BC058278.1 | - | 2 - 3 | 707 | 6528802 | 106 | |
| BC069893.1 | - | 2 - 3 | 715 | 4978765 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|

Egln3
Genomic Map