| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1200013438.mm.1 |
NM_030677.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200013438.mm.1 |
NM_030677.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200013438.mm.1 |
TC1200001955.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200013435.mm.1 |
NM_030677.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200013435.mm.1 |
NM_030677.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200013435.mm.1 |
TC1200001955.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200013439.mm.1 |
NM_030677.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200013439.mm.1 |
NM_030677.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200013439.mm.1 |
TC1200001955.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200013434.mm.1 |
NM_030677.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200013434.mm.1 |
NM_030677.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200013434.mm.1 |
TC1200001955.mm.1 |
MTA TC |