Assay Details
Gene Symbol: Rrm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009104.2 | NP_033130.1 | 2 - 3 | 269 | 85 | |
GenBank mRNA | AK088907.1 | - | 2 - 3 | 239 | 85 | |
AK142027.1 | - | 2 - 3 | 243 | 85 | ||
AK147111.1 | - | 2 - 3 | 236 | 85 | ||
AK161643.1 | - | 2 - 3 | 243 | 85 | ||
AK167078.1 | - | 2 - 3 | 238 | 85 | ||
AK167204.1 | - | 2 - 3 | 241 | 85 | ||
AK168994.1 | - | 2 - 3 | 240 | 85 | ||
BC085136.1 | - | 2 - 3 | 269 | 6402725 | 85 | |
M14223.1 | - | 2 - 3 | 238 | 85 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
