Assay Details
| Gene Symbol: Sag | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009118.2 | NP_033144.1 | 12 - 13 | 1385 | 102 | |
| GenBank mRNA | AK143098.1 | - | 12 - 13 | 1144 | 102 | |
| AK149299.1 | - | 12 - 13 | 1394 | 102 | ||
| BC016498.1 | - | 12 - 13 | 1178 | 4527551 | 102 | |
| M24086.1 | - | 12 - 13 | 1172 | 102 | ||
More Information
Set Membership: |
|

Sag
Genomic Map