Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100012750.mm.1 |
NM_009107.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012750.mm.1 |
NM_009107.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012750.mm.1 |
TC0100001554.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100012751.mm.1 |
NM_009107.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012751.mm.1 |
NM_009107.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012751.mm.1 |
TC0100001554.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100012752.mm.1 |
NM_009107.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012752.mm.1 |
NM_009107.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012752.mm.1 |
TC0100001554.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100012748.mm.1 |
NM_009107.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012748.mm.1 |
NM_009107.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012748.mm.1 |
TC0100001554.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100012754.mm.1 |
NM_009107.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012754.mm.1 |
NM_009107.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012754.mm.1 |
TC0100001554.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100012755.mm.1 |
NM_009107.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012755.mm.1 |
NM_009107.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012755.mm.1 |
TC0100001554.mm.1 |
MTA TC |