Assay Details
| Gene Symbol: Apobec4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081197.1 | NP_001074666.1 | 1 - 2 | 216 | 95 | |
| GenBank mRNA | AK017014.1 | - | 1 - 2 | 191 | 95 | |
| BC100465.1 | - | 1 - 2 | 216 | 95 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC100465.1, NM_001081197.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
mRNA processing
biological_process
molecular_function
GTP cyclohydrolase activity
tRNA-specific adenosine deaminase activity
zinc ion binding
hydrolase activity
hydrolase activity, acting on carbon-nitrogen (but not peptide) bonds, in cyclic amidines
N-cyclopropylmelamine deaminase activity
N-cyclopropylammeline deaminase activity
N-cyclopropylammelide alkylamino hydrolase activity
2,5-diamino-6-ribitylamino-4(3H)-pyrimidinone 5'-phosphate deaminase activity
tRNA-specific adenosine-37 deaminase activity
archaeal-specific GTP cyclohydrolase activity
metal ion binding
tRNA-specific adenosine-34 deaminase activity
mRNA processing
biological_process
molecular_function
GTP cyclohydrolase activity
tRNA-specific adenosine deaminase activity
zinc ion binding
hydrolase activity
hydrolase activity, acting on carbon-nitrogen (but not peptide) bonds, in cyclic amidines
N-cyclopropylmelamine deaminase activity
N-cyclopropylammeline deaminase activity
N-cyclopropylammelide alkylamino hydrolase activity
2,5-diamino-6-ribitylamino-4(3H)-pyrimidinone 5'-phosphate deaminase activity
tRNA-specific adenosine-37 deaminase activity
archaeal-specific GTP cyclohydrolase activity
metal ion binding
tRNA-specific adenosine-34 deaminase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P221035 | AK017014.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P221035 | BC100465.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P221035 | NM_001081197.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P221035 | AK017014.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P221035 | BC100465.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P221035 | NM_001081197.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 943675 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 665408 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 683314 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 4866 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P149129 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P221035 | AK017014.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P221035 | BC100465.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P221035 | NM_001081197.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P221035 | AK017014.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P221035 | BC100465.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P221035 | NM_001081197.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P221035 | AK017014.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P221035 | BC100465.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P221035 | NM_001081197.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P221035 | AK017014.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P221035 | BC100465.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P221035 | NM_001081197.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P120221 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20722740 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20722753 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20722684 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20722745 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20722743 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20722740 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20722709 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20722752 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20722717 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20722707 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00500230 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20722757 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00087925 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20722722 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20722742 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20722744 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20722751 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20722688 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20722746 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20722720 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00287453 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20722712 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00465332 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00326952 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20722694 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20722710 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20722733 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00557546 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20722704 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00718562 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20722729 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20722715 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20722718 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20722748 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20722678 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20722687 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01098698 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20722728 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20722705 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00135414 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20722700 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00579459 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20722692 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20722758 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20722735 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20722736 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20722690 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20722703 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20722713 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20722734 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20722702 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20722680 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20722723 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20722738 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20722681 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20722759 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10350682 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10350683 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1454089_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10350681 | - | - |
| Mouse Genome 430 2.0 Array | 1454089_at | - | - |
| Mouse Genome 430 2.0 Array | 1454089_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71281_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71281_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100011319.mm.1 | NM_001081197.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100011319.mm.1 | NM_001081197.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100011319.mm.1 | TC0100001367.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100011321.mm.1 | NM_001081197.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100011321.mm.1 | NM_001081197.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100011321.mm.1 | TC0100001367.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100011322.mm.1 | NM_001081197.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100011322.mm.1 | NM_001081197.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100011322.mm.1 | TC0100001367.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100011320.mm.1 | NM_001081197.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100011320.mm.1 | NM_001081197.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100011320.mm.1 | TC0100001367.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1454089_PM_at | - | - |
| Mouse Expression Set 430 | 1454089_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1454089_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1454089_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 765018 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1221296 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1221296 | - | - |

Apobec4
Genomic Map