Assay Details
Gene Symbol: Prrx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK008428.2 | - | 1 - 2 | 45 | 53 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK008428.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
