Assay Details
Gene Symbol: 4933436E23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040455.1 | 6 - 7 | 563 | 120 | ||
GenBank mRNA | AK017079.1 | - | 6 - 7 | 563 | 120 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
crumbs family member 1, photoreceptor morphogenesis associated |
Crb1 |
7530426H14Rik, A930008G09Rik |
-19.4kb | Anti-sense | Chr. 1 - 139197546 - 139379118 on NCBI Build GRCm38 |
Downstream |
predicted gene, 34816 |
Gm34816 | 9.0kb | Sense | Chr. 1 - 139408280 - 139413864 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
