Assay Details
| Gene Symbol: Maoa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173740.3 | NP_776101.3 | 12 - 13 | 1408 | 75 | |
| GenBank mRNA | AK004337.1 | - | 12 - 13 | 1256 | 75 | |
| AK078642.1 | - | 5 - 6 | 395 | 75 | ||
| AK146612.1 | - | 12 - 13 | 1409 | 75 | ||
| AK155052.1 | - | 12 - 13 | 1271 | 75 | ||
| AK164457.1 | - | 12 - 13 | 1245 | 75 | ||
| BC029100.1 | - | 12 - 13 | 1278 | 3481702 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Maoa
Genomic Map