Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0X00014728.mm.1 |
NR_033213.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00014728.mm.1 |
TC0X00002382.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00014726.mm.1 |
NR_033213.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00014726.mm.1 |
TC0X00002382.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00014727.mm.1 |
NR_033213.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00014727.mm.1 |
TC0X00002382.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00014730.mm.1 |
NR_033213.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00014730.mm.1 |
TC0X00002382.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00014725.mm.1 |
NR_033213.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00014725.mm.1 |
TC0X00002382.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00014729.mm.1 |
NR_033213.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00014729.mm.1 |
TC0X00002382.mm.1 |
MTA TC |