Assay Details
Gene Symbol: Impa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018864.5 | NP_061352.2 | 4 - 5 | 422 | 65 | |
GenBank mRNA | AF042730.1 | - | 4 - 5 | 367 | 65 | |
AK078406.1 | - | 4 - 5 | 371 | 65 | ||
AK083238.1 | - | 4 - 5 | 356 | 65 | ||
AK145472.1 | - | 359 | 65 | |||
AK147778.1 | - | 4 - 5 | 382 | 65 | ||
AK152774.1 | - | 4 - 5 | 373 | 65 | ||
AK153457.1 | - | 4 - 5 | 389 | 65 | ||
AK160203.1 | - | 4 - 5 | 360 | 65 | ||
AK160467.1 | - | 4 - 5 | 354 | 65 | ||
AK165977.1 | - | 4 - 5 | 364 | 65 | ||
AK168457.1 | - | 4 - 5 | 397 | 65 | ||
BC049080.1 | - | 4 - 5 | 363 | 6419835 | 65 | |
BC055722.1 | - | 3 - 4 | 317 | 6413389 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
