Assay Details
| Gene Symbol: Fabp12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029310.1 | NP_083586.1 | 4 - 5 | 564 | 126 | |
| GenBank mRNA | AK005765.1 | - | 3 - 4 | 410 | 126 | |
| BC030910.1 | - | 4 - 5 | 564 | 4505673 | 126 | |
| EU733649.1 | - | 3 - 4 | 540 | 126 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fabp12
Genomic Map