Assay Details
Gene Symbol: Psmc3ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008949.3 | NP_032975.1 | 6 - 7 | 597 | 97 | |
GenBank mRNA | AB000121.1 | - | 6 - 7 | 562 | 97 | |
AK133462.1 | - | 6 - 7 | 565 | 97 | ||
BC030169.1 | - | 6 - 7 | 574 | 5027493 | 97 | |
FJ937966.1 | - | 6 - 7 | 568 | 97 | ||
FJ937967.1 | - | 5 - 6 | 632 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
