Assay Details
Gene Symbol: Mlx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159384.1 | NP_001152856.1 | 1 - 2 | 82 | 94 | |
NM_001159385.1 | NP_001152857.1 | 1 - 2 | 82 | 94 | ||
GenBank mRNA | AK011478.1 | - | 1 - 2 | 62 | 92 | |
CK620266.1 | - | 1 - 2 | 86 | 94 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001159384.1, NM_001159385.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
