Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100015277.mm.1 |
NM_001159384.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100015277.mm.1 |
NM_001159384.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100015277.mm.1 |
NM_001159385.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100015277.mm.1 |
NM_001159385.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100015277.mm.1 |
NM_011550.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100015277.mm.1 |
NM_011550.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100015277.mm.1 |
TC1100001641.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100015279.mm.1 |
NM_001159384.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100015279.mm.1 |
NM_001159384.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100015279.mm.1 |
NM_001159385.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100015279.mm.1 |
NM_001159385.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100015279.mm.1 |
NM_011550.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100015279.mm.1 |
NM_011550.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100015279.mm.1 |
TC1100001641.mm.1 |
MTA TC |