Assay Details
Gene Symbol: 4930539M17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040597.1 | 1 - 2 | 193 | 132 | ||
GenBank mRNA | AK016002.1 | - | 1 - 2 | 200 | 132 | |
BB615931.1 | - | 1 - 2 | 193 | 132 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | tumor protein D52 | Tpd52 |
AI043038, mD52 |
-57.2kb | Anti-sense | Chr. 3 - 8928626 - 9004769 on NCBI Build GRCm38 |
Downstream | zinc finger and BTB domain containing 10 | Zbtb10 |
4832414A18, AU021107, RINZF |
185.2kb | Sense | Chr. 3 - 9250567 - 9285333 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 104130 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 891934 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 334420 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P105066 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P105066 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139862 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P105066 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02511961 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21935437 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02319140 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02669181 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02299595 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21935430 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21935468 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02660283 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21935431 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02501671 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21935541 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03074037 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21935408 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02734223 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21935528 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02299596 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21935521 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21935422 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21935477 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21935425 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21935480 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21935537 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21935428 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02511957 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02301115 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21935523 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21935407 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21935440 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21935442 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02918836 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21935525 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21935526 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21935487 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21935520 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21935531 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21935417 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02889344 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21935459 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21935539 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02669178 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21935533 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21935416 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21935462 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02412591 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02835463 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02511959 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21935433 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21935543 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21935546 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02518918 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21935544 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21935535 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21935423 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21935545 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21935405 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21935452 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03074039 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21935439 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02747106 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02417157 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21935465 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21935529 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02889337 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02734219 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02847283 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21935489 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21935427 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03148756 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21935457 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Clariom S Array | PSR0300000188.mm.1 | NR_040597.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300000188.mm.1 | NR_040597.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300000188.mm.1 | TC0300000052.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0300000187.mm.1 | NR_040597.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300000187.mm.1 | NR_040597.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300000187.mm.1 | TC0300000052.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0300000181.mm.1 | NR_040597.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300000181.mm.1 | NR_040597.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300000181.mm.1 | TC0300000052.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0300000182.mm.1 | NR_040597.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300000182.mm.1 | NR_040597.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300000182.mm.1 | TC0300000052.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0300000184.mm.1 | NR_040597.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300000184.mm.1 | NR_040597.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300000184.mm.1 | TC0300000052.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0300000186.mm.1 | NR_040597.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300000186.mm.1 | NR_040597.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300000186.mm.1 | TC0300000052.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 487025 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 103780167 | - | - |
