Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300012839.mm.1 |
NM_001163301.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012839.mm.1 |
NM_001163302.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012839.mm.1 |
NM_008994.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012839.mm.1 |
TC0300001601.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300012836.mm.1 |
NM_001163301.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012836.mm.1 |
NM_001163302.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012836.mm.1 |
NM_001163305.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012836.mm.1 |
NM_001163306.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012836.mm.1 |
NM_001267714.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012836.mm.1 |
NM_001267715.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012836.mm.1 |
NM_008994.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012836.mm.1 |
TC0300001601.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300012837.mm.1 |
NM_001163301.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012837.mm.1 |
NM_001163302.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012837.mm.1 |
NM_001163305.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012837.mm.1 |
NM_001163306.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012837.mm.1 |
NM_001267714.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012837.mm.1 |
NM_001267715.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012837.mm.1 |
NM_008994.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012837.mm.1 |
TC0300001601.mm.1 |
MTA TC |