Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100025051.mm.1 |
NM_178626.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100025051.mm.1 |
NM_178626.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100025051.mm.1 |
TC1100002766.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100025054.mm.1 |
NM_178626.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100025054.mm.1 |
NM_178626.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100025054.mm.1 |
TC1100002766.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100025058.mm.1 |
NM_178626.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100025058.mm.1 |
NM_178626.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100025058.mm.1 |
TC1100002766.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100025048.mm.1 |
NM_178626.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100025048.mm.1 |
NM_178626.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100025048.mm.1 |
TC1100002766.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100025047.mm.1 |
NM_178626.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100025047.mm.1 |
NM_178626.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100025047.mm.1 |
TC1100002766.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100025053.mm.1 |
NM_178626.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100025053.mm.1 |
NM_178626.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100025053.mm.1 |
TC1100002766.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100025057.mm.1 |
NM_178626.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100025057.mm.1 |
NM_178626.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100025057.mm.1 |
TC1100002766.mm.1 |
MTA TC |