Assay Details
| Gene Symbol: Kif3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290805.1 | NP_001277734.1 | 2 - 3 | 444 | 154 | |
| NM_001290806.1 | NP_001277735.1 | 2 - 3 | 444 | 154 | ||
| NM_008443.4 | NP_032469.2 | 2 - 3 | 444 | 154 | ||
| GenBank mRNA | AK147078.1 | - | 2 - 3 | 394 | 154 | |
| AK161223.1 | - | 2 - 3 | 435 | 154 | ||
| AK161920.1 | - | 2 - 3 | 418 | 154 | ||
| AK169425.1 | - | 2 - 3 | 418 | 154 | ||
| BC023936.1 | - | 2 - 3 | 394 | 154 | ||
| BC044720.1 | - | 2 - 3 | 405 | 154 | ||
| BC052707.1 | - | 2 - 3 | 410 | 5702415 | 154 | |
| D12645.1 | - | 2 - 3 | 420 | 154 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kif3a
Genomic Map