Assay Details
| Gene Symbol: Slc22a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019687.3 | NP_062661.1 | 2 - 3 | 616 | 63 | |
| GenBank mRNA | AB016257.1 | - | 2 - 3 | 623 | 63 | |
| BC010590.1 | - | 2 - 3 | 616 | 3670138 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc22a4
Genomic Map