Assay Details
| Gene Symbol: Slc22a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011396.3 | NP_035526.1 | 7 - 8 | 1426 | 63 | |
| GenBank mRNA | AB015800.1 | - | 7 - 8 | 1321 | 63 | |
| AF110417.1 | - | 7 - 8 | 1374 | 63 | ||
| AF111425.1 | - | 7 - 8 | 1361 | 63 | ||
| AK140640.1 | - | 7 - 8 | 1422 | 63 | ||
| AK167451.1 | - | 7 - 8 | 1402 | 63 | ||
| BC031118.1 | - | 7 - 8 | 1383 | 4981427 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc22a5
Genomic Map