Assay Details
| Gene Symbol: P4ha2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136076.2 | NP_001129548.1 | 4 - 5 | 451 | 54 | |
| NM_011031.2 | NP_035161.2 | 4 - 5 | 451 | 54 | ||
| GenBank mRNA | AK053504.1 | - | 4 - 5 | 412 | 54 | |
| AK142624.1 | - | 4 - 5 | 400 | 54 | ||
| AK144124.1 | - | 4 - 5 | 397 | 54 | ||
| AK145535.1 | - | 4 - 5 | 378 | 54 | ||
| BC018411.1 | - | 4 - 5 | 363 | 3157481 | 54 | |
| U16163.1 | - | 3 - 4 | 338 | 54 | ||
More Information
Updated Mapping Information
Set Membership: |
|

P4ha2
Genomic Map