Assay Details
| Gene Symbol: P4ha2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136076.2 | NP_001129548.1 | 9 - 10 | 1354 | 96 | |
| NM_011031.2 | NP_035161.2 | 9 - 10 | 1354 | 96 | ||
| GenBank mRNA | AK053504.1 | - | 9 - 10 | 1314 | 96 | |
| AK142689.1 | - | 1 - 2 | 49 | 96 | ||
| AK144124.1 | - | 9 - 10 | 1301 | 96 | ||
| AK145535.1 | - | 9 - 10 | 1281 | 96 | ||
| BC018411.1 | - | 9 - 10 | 1266 | 3157481 | 96 | |
| U16163.1 | - | 8 - 9 | 1241 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

P4ha2
Genomic Map