Assay Details
| Gene Symbol: Vdac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011694.4 | NP_035824.1 | 1 - 2 | 42 | 61 | |
| GenBank mRNA | AK002740.1 | - | 1 - 2 | 103 | 61 | |
| AK146239.1 | - | 1 - 2 | 75 | 61 | ||
| AK153094.1 | - | 1 - 2 | 44 | 61 | ||
| AK167860.1 | - | 1 - 2 | 75 | 61 | ||
| AK168225.1 | - | 1 - 2 | 51 | 61 | ||
| AK168672.1 | - | 1 - 2 | 80 | 61 | ||
| AK169160.1 | - | 1 - 2 | 80 | 61 | ||
| AK169282.1 | - | 1 - 2 | 84 | 61 | ||
| AK169354.1 | - | 1 - 2 | 80 | 61 | ||
| AK169671.1 | - | 1 - 2 | 41 | 61 | ||
| U30840.1 | - | 1 - 2 | 42 | 61 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK146239.1, AK153094.1, AK167860.1, AK168225.1, AK168672.1, AK169160.1, AK169282.1, AK169354.1, AK169671.1, NM_011694.4, U30840.1 transcripts.
Set Membership: |
|

Vdac1
Genomic Map