Assay Details
Gene Symbol: Cnot6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290741.1 | NP_001277670.1 | 1 - 2 | 116 | 67 | |
NM_212484.2 | NP_997649.1 | 2 - 3 | 485 | 67 | ||
GenBank mRNA | AK187687.1 | - | 2 - 3 | 132 | 67 | |
AY043269.1 | - | 1 - 2 | 114 | 67 | ||
BC062950.1 | - | 2 - 3 | 473 | 6840162 | 67 | |
BY741392.1 | - | 2 - 3 | 487 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
