Assay Details
Gene Symbol: Cnot6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212484.2 | NP_997649.1 | 5 - 6 | 843 | 95 | |
GenBank mRNA | AK217278.1 | - | 3 - 4 | 204 | 95 | |
BC062950.1 | - | 5 - 6 | 831 | 6840162 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
