Assay Details
Gene Symbol: Cnot6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290741.1 | NP_001277670.1 | 9 - 10 | 1258 | 74 | |
NM_212484.2 | NP_997649.1 | 10 - 11 | 1612 | 74 | ||
GenBank mRNA | AK129307.1 | - | 4 - 5 | 2550 | 74 | |
AK212064.1 | - | 1 - 2 | 96 | 74 | ||
AK213936.1 | - | 1 - 2 | 85 | 74 | ||
AY043269.1 | - | 9 - 10 | 1256 | 74 | ||
BC023766.1 | - | 5 - 6 | 723 | 74 | ||
BC049984.1 | - | 5 - 6 | 782 | 74 | ||
BC057190.1 | - | 4 - 5 | 709 | 74 | ||
BC062950.1 | - | 10 - 11 | 1600 | 6840162 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
