Assay Details
| Gene Symbol: Ltc4s | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008521.1 | NP_032547.1 | 2 - 3 | 196 | 62 | |
| GenBank mRNA | BC028760.1 | - | 2 - 3 | 196 | 1348372 | 62 |
| BC038518.1 | - | 2 - 3 | 206 | 62 | ||
| U27195.1 | - | 2 - 3 | 196 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ltc4s
Genomic Map