Assay Details
| Gene Symbol: Ltc4s | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC028760.1 | - | 4 - 5 | 349 | 1348372 | 109 |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ltc4s| Gene Symbol: Ltc4s | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC028760.1 | - | 4 - 5 | 349 | 1348372 | 109 |
Set Membership: |
|