Assay Details
| Gene Symbol: Zfp354a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC087540.1 | - | 2 - 3 | 362 | 4239789 | 63 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC087540.1 transcripts.
Set Membership: |
|

Zfp354a
Genomic Map