Assay Details
Gene Symbol: Thg1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK007233.1 | - | 2 - 3 | 101 | 69 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK007233.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
