Assay Details
Gene Symbol: Thg1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290737.1 | NP_001277666.1 | 2 - 3 | 168 | 93 | |
GenBank mRNA | AK007233.1 | - | 3 - 4 | 206 | 93 | |
AK016076.1 | - | 2 - 3 | 169 | 93 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK007233.1, AK016076.1, NM_001290737.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
