Assay Details
Gene Symbol: Thg1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080969.3 | NP_001074438.1 | 2 - 3 | 400 | 64 | |
NM_001290737.1 | NP_001277666.1 | 3 - 4 | 343 | 64 | ||
GenBank mRNA | AK007233.1 | - | 4 - 5 | 381 | 64 | |
AK010876.1 | - | 2 - 3 | 392 | 64 | ||
AK015467.1 | - | 2 - 3 | 194 | 64 | ||
AK016076.1 | - | 3 - 4 | 344 | 64 | ||
AK017532.1 | - | 2 - 3 | 383 | 64 | ||
BC092541.1 | - | 2 - 3 | 380 | 30280958 | 64 | |
BC115560.1 | - | 2 - 3 | 388 | 40101901 | 64 | |
BC115561.1 | - | 2 - 3 | 388 | 40101902 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
