Assay Details
Gene Symbol: Thg1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290737.1 | NP_001277666.1 | 1 - 2 | 68 | 73 | |
GenBank mRNA | AK016076.1 | - | 1 - 2 | 69 | 73 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK016076.1, NM_001290737.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
