Assay Details
| Gene Symbol: Atp10b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176999.3 | NP_795973.2 | 19 - 20 | 3658 | 67 | |
| GenBank mRNA | AK220352.1 | - | 9 - 10 | 1804 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atp10b
Genomic Map