Assay Details
| Gene Symbol: Ikbke | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019777.3 | NP_062751.2 | 15 - 16 | 1978 | 102 | |
| GenBank mRNA | AB016589.1 | - | 13 - 14 | 1666 | 102 | |
| AK088580.1 | - | 15 - 16 | 1980 | 102 | ||
| AK149911.1 | - | 15 - 16 | 1967 | 102 | ||
| BC037446.1 | - | 14 - 15 | 1650 | 3980145 | 102 | |
| CT010206.1 | - | 13 - 14 | 1632 | 102 | ||
| CT010313.1 | - | 13 - 14 | 1560 | 102 | ||
More Information
Set Membership: |
|

Ikbke
Genomic Map