Assay Details
Gene Symbol: Cpb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029706.1 | NP_083982.1 | 2 - 3 | 221 | 99 | |
GenBank mRNA | AK003061.1 | - | 2 - 3 | 160 | 99 | |
BC138759.1 | - | 2 - 3 | 179 | 8861781 | 99 | |
BC138762.1 | - | 2 - 3 | 179 | 8861784 | 99 | |
BQ831426.1 | - | 2 - 3 | 237 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
